Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210404O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256493.1 | NP_001243422.1 | 5 | 1535 | ||
GenBank mRNA | BC089531.1 | - | 1 | 2043 | ||
BC141011.1 | - | 4 | 1058 | 9055623 |