Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197959.2 | NP_932063.2 | 13 | 1987 | ||
GenBank mRNA | AK013867.1 | - | 2 | 190 | ||
AK156592.1 | - | 13 | 1957 | |||
BC023289.1 | - | 7 | 948 | |||
BC049272.1 | - | 4 | 521 | |||
BC057614.1 | - | 13 | 1955 | 6401536 |