Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172404.2 | NP_765992.2 | 6 | 653 | ||
GenBank mRNA | AK008165.1 | - | 4 | 430 | ||
AK031511.1 | - | 6 | 666 | |||
AK042391.1 | - | 5 | 599 | |||
AK088404.1 | - | 5 | 561 | |||
AK163165.1 | - | 5 | 462 | |||
BC016206.1 | - | 6 | 755 | 4912335 | ||
BC025124.1 | - | 5 | 786 | 4486058 | ||
BC052047.1 | - | 6 | 653 | 5687475 |