Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027376.3 | NP_081652.2 | 8, 9 | 1062 | ||
GenBank mRNA | AK033247.1 | - | 8, 9 | 1049 | ||
AK045745.1 | - | 1057 | ||||
AK134650.1 | - | 8, 9 | 1062 | |||
CK624547.1 | - | 3, 4 | 324 |