Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005940.4 | NP_005931.2 | 5 | 764 | ||
NR_133013.1 | 5 | 716 | ||||
GenBank mRNA | AK075448.1 | - | 5 | 686 | ||
AK125911.1 | - | 5 | 2296 | |||
AK129792.1 | - | 5 | 734 | |||
BC057788.1 | - | 5 | 713 | 30348138 | ||
X57766.1 | - | 5 | 721 |