Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027407.3 | NP_081683.3 | 7 | 1000 | ||
GenBank mRNA | AK006525.1 | - | 5 | 528 | ||
AK029991.1 | - | 7 | 772 | |||
AK158859.1 | - | 7 | 836 | |||
BC058983.1 | - | 2 | 129 | |||
BC079669.1 | - | 7 | 1000 | 30355976 |