Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054042.2 | NP_473383.1 | 1 | 2233 | ||
GenBank mRNA | AF378758.1 | - | 1 | 2191 | ||
AF388572.1 | - | 1 | 2188 | |||
AK011844.1 | - | 1 | 1059 | |||
BC046318.1 | - | 1 | 2233 | 6314798 | ||
BC128295.1 | - | 1 | 330 | |||
BC152362.1 | - | 1 | 330 |