Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027430.2 | NP_081706.1 | 4 | 484 | ||
GenBank mRNA | AK002233.2 | - | 4 | 433 | ||
AK011889.1 | - | 4 | 489 | |||
AK151522.1 | - | 4 | 471 | |||
BC018324.1 | - | 4 | 330 | 3501265 |