Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 3 | 457 | ||
GenBank mRNA | AK012014.1 | - | 3 | 448 | ||
AK048488.1 | - | 3 | 480 | |||
AK050391.1 | - | 3 | 457 | |||
AK078856.1 | - | 4 | 416 | |||
AK131608.1 | - | 2 | 368 | |||
AK161812.1 | - | 3 | 432 | |||
AK163839.1 | - | 452 | ||||
AK168006.1 | - | 3 | 459 | |||
AK168752.1 | - | 3 | 479 | |||
AK169128.1 | - | 3 | 458 | |||
AK191398.1 | - | 2 | 175 | |||
BC005755.1 | - | 3 | 412 | 2654151 |