Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172064.1 | NP_001165535.1 | 8 | 1222 | ||
NM_001172065.1 | NP_001165536.1 | 8 | 1222 | |||
NM_153777.2 | NP_722472.1 | 8 | 808 | |||
GenBank mRNA | AK017603.1 | - | 8 | 1196 | ||
AK220237.1 | - | 8 | 1176 | |||
BC027807.1 | - | 8 | 818 | 5324022 |