Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027475.3 | NP_081751.1 | 12 | 1772 | ||
GenBank mRNA | AK017681.1 | - | 10 | 1196 | ||
AK028664.1 | - | 12 | 1736 | |||
AK043261.1 | - | 12 | 1621 | |||
BC013470.1 | - | 12 | 1721 | 4220504 |