Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027480.3 | NP_081756.4 | 17 | 1860 | ||
GenBank mRNA | AK019475.1 | - | 4 | 526 | ||
AK043638.1 | - | 15 | 2942 | |||
AK173316.1 | - | 15 | 2826 | |||
BC017633.1 | - | 4 | 531 | |||
BC048236.1 | - | 16 | 1765 | 6313999 |