Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109985.1 | NP_001103455.1 | 5 | 832 | ||
NM_027528.2 | NP_081804.1 | 5 | 649 | |||
GenBank mRNA | AK018149.1 | - | 5 | 832 | ||
AK138471.1 | - | 5 | 650 | |||
AK162919.1 | - | 5 | 650 |