Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027665.1 | NP_081941.1 | 1 | 1620 | ||
GenBank mRNA | AK077058.1 | - | 1773 | |||
AY190760.1 | - | 1 | 1620 | |||
BC113152.1 | - | 1 | 1612 | 40059344 | ||
BC113762.1 | - | 1 | 1612 | 40059342 |