Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027732.2 | NP_082008.1 | 6, 7 | 895 | ||
GenBank mRNA | AF542046.1 | - | 6, 7 | 892 | ||
AK017020.1 | - | 6, 7 | 893 | |||
BC100434.1 | - | 6, 7 | 895 | 30918218 | ||
BC138203.1 | - | 6, 7 | 826 | 8861222 | ||
BC145988.1 | - | 6, 7 | 826 | 40131291 |