Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153196.1 | NP_694876.1 | 4 | 343 | ||
GenBank mRNA | AK017229.1 | - | 4 | 361 | ||
AK138939.1 | - | 4 | 361 | |||
BC023339.1 | - | 4 | 343 | 5253827 |