Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290521.1 | NP_001277450.1 | 10 | 1212 | ||
NM_001290522.1 | NP_001277451.1 | 10 | 1102 | |||
NM_023597.2 | NP_076086.1 | 10 | 1105 | |||
GenBank mRNA | AF274321.1 | - | 10 | 1082 | ||
AK017259.1 | - | 10 | 1095 | |||
AK141026.1 | - | 10 | 1210 | |||
BC026500.1 | - | 9 | 991 | |||
BC037102.1 | - | 10 | 1095 | 5364307 |