Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168371.2 | NP_001161843.1 | 16 | 2481 | ||
NM_001168372.2 | NP_001161844.1 | 16 | 2478 | |||
NM_001168373.2 | NP_001161845.1 | 16 | 2475 | |||
NM_027804.4 | NP_082080.3 | 16 | 2478 | |||
NM_145407.3 | NP_663382.2 | 15 | 2406 | |||
GenBank mRNA | AK043088.1 | - | 15 | 2242 | ||
AK122396.2 | - | 15 | 2264 | |||
AK144099.1 | - | 11 | 1510 | |||
AK146504.1 | - | 16 | 2304 | |||
AK148412.1 | - | 11 | 1467 | |||
AK159473.1 | - | 16 | 2300 | |||
AK160807.1 | - | 16 | 2304 | |||
AK171942.1 | - | 16 | 2307 | |||
AK183398.1 | - | 2 | 100 | |||
BC006916.1 | - | 8 | 1012 | |||
BC046824.1 | - | 7 | 941 | |||
BC060613.1 | - | 16 | 2406 | 6400986 |