Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027840.3 | NP_082116.1 | 3 | 408 | ||
GenBank mRNA | AF395844.1 | - | 2 | 247 | ||
AK018632.1 | - | 3 | 410 | |||
AK042068.1 | - | 3 | 383 | |||
AK157032.1 | - | 3 | 381 | |||
AK157884.1 | - | 3 | 382 | |||
BC039809.1 | - | 3 | 314 | 2938247 |