Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163579.1 | NP_001157051.1 | 13 | 2129 | ||
NM_001163580.1 | NP_001157052.1 | 13 | 2081 | |||
NM_028915.3 | NP_083191.2 | 13 | 2129 | |||
GenBank mRNA | AK004646.1 | - | 13 | 2130 | ||
AK016559.1 | - | 13 | 2068 | |||
AK029563.1 | - | 13 | 2047 | |||
AK030800.1 | - | 5 | 723 | |||
AK078433.1 | - | 2 | 124 | |||
AK173256.1 | - | 13 | 2057 | |||
BC061202.1 | - | 13 | 2037 | |||
BC138734.1 | - | 13 | 2103 | 8861756 | ||
BC138735.1 | - | 13 | 2103 | 8861757 |