Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143776.1 | NP_001137248.1 | 8 | 1069 | ||
NM_001143777.1 | NP_001137249.1 | 9 | 1173 | |||
NM_024244.4 | NP_077206.3 | 8 | 1069 | |||
GenBank mRNA | AK004797.1 | - | 8 | 1053 | ||
AK032053.1 | - | 9 | 985 | |||
AK036299.1 | - | 9 | 1136 | |||
AK041147.1 | - | 8 | 1070 | |||
AK044049.1 | - | 9 | 1096 | |||
AK082202.1 | - | 8 | 1065 | |||
AK220421.1 | - | 9 | 1009 | |||
BC129839.1 | - | 8 | 1025 | 40060810 | ||
BC129840.1 | - | 8 | 1025 | 40060811 |