Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023626.4 | NP_076115.3 | 3 | 267 | ||
GenBank mRNA | AK030364.1 | - | 2 | 74 | ||
AK080787.1 | - | 3 | 236 | |||
AK131993.1 | - | 3 | 267 | |||
AK146145.1 | - | 2 | 84 | |||
AY007791.1 | - | 3 | 236 | |||
BC018342.1 | - | 3 | 217 | 4010253 |