Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170591.1 | NP_733479.1 | 3 | 376 | ||
GenBank mRNA | AK006062.1 | - | 3 | 478 | ||
AK029688.1 | - | 430 | ||||
AK078279.1 | - | 3 | 424 | |||
AK086412.1 | - | 3 | 444 | |||
AK129134.1 | - | 3 | 413 | |||
AK133721.1 | - | 3 | 387 | |||
AK139895.1 | - | 3 | 506 | |||
AK146477.1 | - | 3 | 376 | |||
AK163792.1 | - | 3 | 405 | |||
AK194884.1 | - | 3 | 190 | |||
BC026743.1 | - | 3 | 407 | 3964978 |