Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310003H01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027980.2 | NP_082256.2 | 3 | 960 | ||
GenBank mRNA | AK009114.1 | - | 3 | 924 | ||
AK082976.1 | - | 3 | 961 | |||
AK147900.1 | - | 1047 | ||||
AK148234.1 | - | 3 | 945 | |||
AK165631.1 | - | 2 | 831 | |||
BC034203.1 | - | 3 | 831 | 4986832 |