Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122683.1 | NP_001116155.1 | 2 | 333 | ||
NM_175177.4 | NP_780386.3 | 2 | 314 | |||
GenBank mRNA | AK009575.1 | - | 2 | 289 | ||
AK076718.1 | - | 2 | 287 | |||
AK137955.1 | - | 2 | 274 | |||
AK145711.1 | - | 2 | 334 | |||
AK146321.1 | - | 2 | 338 | |||
AK146962.1 | - | 2 | 295 | |||
BC043683.1 | - | 2 | 317 | 5133856 | ||
BC096457.1 | - | 2 | 262 | 4975979 | ||
BY148101.1 | - | 2 | 314 |