Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028011.2 | NP_082287.1 | 4 | 338 | ||
GenBank mRNA | AF395837.1 | - | 4 | 302 | ||
AJ505007.1 | - | 4 | 299 | |||
AK009826.1 | - | 4 | 301 | |||
AK033712.1 | - | 4 | 340 | |||
AK083630.1 | - | 300 |