Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028018.2 | NP_082294.1 | 3 | 384 | ||
GenBank mRNA | AK010262.1 | - | 3 | 354 | ||
AK050468.1 | - | 3 | 351 | |||
AK088192.1 | - | 3 | 339 | |||
AK122215.1 | - | 1 | 158 | |||
AK152830.1 | - | 3 | 351 | |||
AK161899.1 | - | 3 | 340 | |||
AK207302.1 | - | 1 | 78 | |||
BC017605.1 | - | 3 | 350 | 4507347 | ||
BC039052.1 | - | 3 | 327 | 2631157 |