Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 7 | 1118 | ||
GenBank mRNA | AK005460.1 | - | 7 | 1120 | ||
AK051121.1 | - | 6 | 1336 | |||
AK051628.1 | - | 7 | 1136 | |||
AK081672.1 | - | 7 | 1033 | |||
AK159555.1 | - | 7 | 1163 | |||
AK159871.1 | - | 7 | 1148 | |||
AK166578.1 | - | 7 | 1126 | |||
AK167201.1 | - | 7 | 1139 | |||
AK167613.1 | - | 7 | 1136 | |||
AK169021.1 | - | 7 | 1143 | |||
AK169098.1 | - | 7 | 1163 | |||
AK169410.1 | - | 7 | 1093 | |||
AK190364.1 | - | 1 | 66 | |||
AK216244.1 | - | 1 | 66 | |||
BC005788.1 | - | 3 | 445 | |||
BC039643.1 | - | 3 | 513 | |||
BC055711.1 | - | 7 | 1119 | 6415164 | ||
BC085268.1 | - | 7 | 896 | 6487559 |