Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF361644.1 | - | 2, 3 | 372 | ||
AK005532.1 | - | 2, 3 | 355 | |||
AK031742.1 | - | 2, 3 | 210 | |||
AK049358.1 | - | 2, 3 | 358 | |||
AK085617.1 | - | 2, 3 | 365 | |||
AK086940.1 | - | 2, 3 | 305 | |||
AK143428.1 | - | 2, 3 | 348 | |||
AK220209.1 | - | 2, 3 | 182 | |||
BB857917.1 | - | 2, 3 | 375 | |||
BC013549.1 | - | 2, 3 | 374 | 4222133 | ||
BY295462.1 | - | 2, 3 | 373 |