Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023651.4 | NP_076140.2 | 2 | 646 | ||
GenBank mRNA | AF329877.2 | - | 2 | 555 | ||
AJ304506.1 | - | 2 | 579 | |||
AK011355.1 | - | 2 | 616 | |||
AK029099.1 | - | 2 | 616 | |||
AK029336.1 | - | 2 | 619 | |||
AK031710.1 | - | 2 | 646 | |||
AK031975.1 | - | 2 | 602 | |||
AK032650.1 | - | 1 | 15 | |||
AK037872.1 | - | 1 | 162 | |||
AK153438.1 | - | 2 | 595 | |||
AK169148.1 | - | 2 | 671 | |||
BC023683.1 | - | 2 | 658 | 5343438 |