Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023651.4 | NP_076140.2 | 2 | 211 | ||
GenBank mRNA | AF329877.2 | - | 2 | 120 | ||
AJ304506.1 | - | 2 | 144 | |||
AK011355.1 | - | 2 | 181 | |||
AK029099.1 | - | 2 | 181 | |||
AK029336.1 | - | 2 | 184 | |||
AK031710.1 | - | 2 | 211 | |||
AK153438.1 | - | 2 | 160 | |||
AK169148.1 | - | 2 | 236 | |||
BC023683.1 | - | 2 | 223 | 5343438 |