Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028115.3 | NP_082391.1 | 4 | 499 | ||
NM_029839.3 | NP_084115.2 | 4 | 499 | |||
GenBank mRNA | AK011362.1 | - | 4 | 494 | ||
AK018525.1 | - | 4 | 472 | |||
AK031674.1 | - | 4 | 477 | |||
AK164095.1 | - | 4 | 470 | |||
AK169221.1 | - | 4 | 465 | |||
BC039287.1 | - | 4 | 444 | |||
BC054088.1 | - | 4 | 313 | |||
BC089592.1 | - | 4 | 466 | 6757213 |