Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172882.3 | NP_766470.2 | 42 | 7409 | ||
GenBank mRNA | AY336569.1 | - | 42 | 7409 |