Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291070.1 | NP_001277999.1 | 11 | 1203 | ||
NM_175178.4 | NP_780387.2 | 13 | 1609 | |||
GenBank mRNA | AK012958.1 | - | 9 | 961 | ||
AK049928.1 | - | 13 | 1440 | |||
AK158809.1 | - | 12 | 1271 | |||
BC046517.1 | - | 13 | 1595 | |||
BC096476.1 | - | 12 | 1384 | 4221224 | ||
BC120685.1 | - | 12 | 1238 | 40129608 |