Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028149.1 | NP_082425.1 | 7 | 671 | ||
GenBank mRNA | AK012109.1 | - | 7 | 395 | ||
AK144786.1 | - | 7 | 645 | |||
AK207539.1 | - | 3 | 175 | |||
AK220232.1 | - | 7 | 624 | |||
EF649694.1 | - | 7 | 418 |