Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028182.1 | NP_082458.1 | 3, 4 | 889 | ||
GenBank mRNA | AK008803.1 | - | 3, 4 | 675 | ||
AK157396.1 | - | 4, 5 | 687 | |||
AX776203.1 | - | 2, 3 | 346 | |||
BC023784.1 | - | 3, 4 | 282 | |||
BC116682.1 | - | 3, 4 | 597 | 40106612 |