Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033528.1 | NP_001028700.1 | 3 | 401 | ||
GenBank mRNA | AK028572.1 | - | 3 | 446 | ||
AK079342.1 | - | 3 | 286 | |||
AK129363.1 | - | 3 | 199 | |||
AK165635.1 | - | 3 | 349 |