Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081349.2 | NP_001074818.2 | 6 | 777 | ||
NM_001083809.1 | NP_001077278.1 | 5 | 716 | |||
NM_024497.1 | NP_078773.1 | 6 | 675 | |||
GenBank mRNA | AB103034.1 | - | 6 | 746 | ||
AK011417.1 | - | 6 | 775 | |||
AK031215.1 | - | 5 | 719 | |||
AK046758.1 | - | 6 | 769 | |||
AK165374.1 | - | 6 | 769 | |||
BC053747.1 | - | 6 | 574 | 6330614 |