Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081349.2 | NP_001074818.2 | 8 | 1008 | ||
NM_001083809.1 | NP_001077278.1 | 7 | 947 | |||
NM_024497.1 | NP_078773.1 | 8 | 906 | |||
GenBank mRNA | AB103034.1 | - | 8 | 977 | ||
AK011417.1 | - | 8 | 1006 | |||
AK031215.1 | - | 7 | 950 | |||
AK046758.1 | - | 8 | 1000 | |||
AK165374.1 | - | 8 | 1000 | |||
AK214626.1 | - | 3 | 180 | |||
AK216445.1 | - | 3 | 180 | |||
BC053747.1 | - | 8 | 805 | 6330614 |