Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 9 | 1588 | ||
GenBank mRNA | AK011892.1 | - | 5 | 519 | ||
AK046669.1 | - | 9 | 1554 | |||
AK144950.1 | - | 2661 | ||||
AK173311.1 | - | 7 | 3546 | |||
BC031392.1 | - | 9 | 1532 | 4952407 |