Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028250.3 | NP_082526.2 | 5 | 1065 | ||
GenBank mRNA | AK011781.1 | - | 5 | 1058 | ||
AK167959.1 | - | 6 | 1005 | |||
BC115580.1 | - | 5 | 657 | 40102241 | ||
BC115581.1 | - | 5 | 657 | 40102242 |