siRNA Details
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114334.1 | NP_001107806.1 | 3 | 304 | ||
NM_028259.4 | NP_082535.1 | 3 | 304 | |||
GenBank mRNA | AK032724.1 | - | 3 | 266 | ||
AK085687.1 | - | 3 | 273 | |||
AK135716.1 | - | 3 | 272 | |||
AK163258.1 | - | 3 | 274 | |||
AK178169.1 | - | 3 | 186 | |||
BC038491.1 | - | 3 | 277 | 4010710 | ||
BC071244.1 | - | 3 | 261 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
