Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163538.1 | NP_001157010.1 | 2 | 267 | ||
NM_028273.3 | NP_082549.2 | 2 | 267 | |||
GenBank mRNA | AK012159.1 | - | 2 | 243 | ||
AK034588.1 | - | 2 | 251 | |||
AK049246.1 | - | 2 | 227 | |||
AK146216.1 | - | 2 | 268 | |||
AK162266.1 | - | 2 | 218 | |||
AK169643.1 | - | 2 | 230 | |||
BC021317.1 | - | 2 | 217 | |||
BC052179.1 | - | 2 | 191 | |||
BC138924.1 | - | 2 | 250 | 8861946 | ||
BC138925.1 | - | 2 | 250 | 8861947 | ||
BC145467.1 | - | 2 | 250 | 9054029 | ||
BY101410.1 | - | 2 | 281 |