Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028276.1 | NP_082552.1 | 5 | 332 | ||
GenBank mRNA | AK012492.1 | - | 5 | 372 | ||
AK134348.1 | - | 5 | 390 | |||
AK134784.1 | - | 5 | 357 | |||
AK165575.1 | - | 5 | 373 | |||
AK165628.1 | - | 5 | 373 | |||
AK172924.1 | - | 5 | 332 | |||
AY316163.1 | - | 5 | 355 | |||
BC062165.1 | - | 5 | 381 | |||
BC082590.1 | - | 5 | 358 | 30625504 |