Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zufsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028287.2 | NP_082563.1 | 10 | 1758 | ||
GenBank mRNA | AK046191.1 | - | 10 | 1761 | ||
AK052071.1 | - | 10 | 1742 | |||
AK077304.1 | - | 10 | 1806 | |||
AK077730.1 | - | 10 | 1777 | |||
AK160928.1 | - | 10 | 1757 | |||
AK164346.1 | - | 7 | 3233 | |||
AK164710.1 | - | 7 | 3104 | |||
AK169467.1 | - | 3 | 420 | |||
BC067065.1 | - | 8 | 1503 | |||
BC089514.1 | - | 10 | 1771 | 30252639 |