Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028296.3 | NP_082572.1 | 7 | 1598 | ||
GenBank mRNA | AB080741.1 | - | 8 | 1554 | ||
AK132222.1 | - | 7 | 1439 | |||
BC017606.1 | - | 7 | 790 | 4507552 | ||
BC066111.1 | - | 8 | 1482 | 30360743 |