Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133747.2 | NP_598508.2 | 8 | 1473 | ||
GenBank mRNA | AK046481.1 | - | 7 | 1475 | ||
AK146215.1 | - | 8 | 1417 | |||
AK146761.1 | - | 8 | 1439 | |||
AK146955.1 | - | 8 | 1386 | |||
AK167224.1 | - | 8 | 1357 | |||
BC004626.1 | - | 8 | 1370 | 3585698 |