Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145778.1 | NP_001139250.1 | 6 | 1702 | ||
NM_023685.4 | NP_076174.3 | 6 | 1699 | |||
GenBank mRNA | AK013249.1 | - | 1 | 764 | ||
AK028902.1 | - | 6 | 1689 | |||
AK031080.1 | - | 6 | 1660 | |||
AK031286.1 | - | 6 | 1657 | |||
BC007473.1 | - | 6 | 1657 | 3496252 |