Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278415.1 | NP_001265344.1 | 8 | 793 | ||
NM_028388.3 | NP_082664.1 | 7 | 696 | |||
GenBank mRNA | AK013511.1 | - | 7 | 647 | ||
AK078351.1 | - | 7 | 640 | |||
AK088193.1 | - | 4 | 435 | |||
AK146998.1 | - | 7 | 630 | |||
AK150404.1 | - | 8 | 730 | |||
AK151729.1 | - | 8 | 730 | |||
AK159460.1 | - | 7 | 613 | |||
AK166539.1 | - | 7 | 657 | |||
AK169143.1 | - | 7 | 667 | |||
AK185171.1 | - | 3 | 320 | |||
BC030946.1 | - | 7 | 625 | 5003524 | ||
CF106105.1 | - | 6 | 538 |