Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027045.1 | NP_081321.1 | 11 | 2845 | ||
NM_028407.3 | NP_082683.2 | 8 | 932 | |||
GenBank mRNA | AK005931.1 | - | 8 | 932 | ||
AK147383.1 | - | 11 | 2840 | |||
BC111465.1 | - | 9 | 1141 | |||
BC139090.1 | - | 10 | 2633 | 8862112 |